Order Entry
Puerto Rico
ContactUsLinkComponent
Multispecies HSP90aA1 ELISA Kit
Multispecies HSP90aA1 ELISA Kit
Catalog # MSPP-SED523MI
CAS Number:  
undefined
Multispecies HSP90aA1 ELISA Kit
Catalog # MSPP-SED523MI
Supplier Number:  SED523MI
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
    Rat
    Mouse
  • Target protein:
    HSP90aA1
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Heat Shock Protein 90kDa Alpha A1 (HSP90aA1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    1.56 - 100 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.65 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SED523MI
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human/Mouse/Rat HSP90aA1 (Heat Shock Protein 90kDa Alpha A1). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.65 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1) and mitochondria (TRAP1). In humans, 2 distinct HSP90 cDNA clones were identified that hybrid-select different mRNA transcripts encoding 2 members of the HSP90 family. These were called HSP89-alpha and HSP89-beta

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.