- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:HumanRatMouse
- Target protein:H2A
- Size:1 Kit
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Histone H2A (H2A) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:0.312 - 20 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.117 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA282MI
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human/Mouse/Rat H2A (Histone H2A). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Histone H2A is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the 'beads on a string' structure.The term 'Histone H2A' is intentionally non-specific and refers to a variety of closely related proteins that vary often by only a few amino acids. Notable variants include H2A.1, H2A.2, H2A.X, and H2A.Z. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is located on chromosome 12 and encodes a variant H2A histone.