Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse UCP1 ELISA Kit
Mouse UCP1 ELISA Kit
Catalog # MSPP-SEF557MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse UCP1 ELISA Kit
Catalog # MSPP-SEF557MU
Supplier:  Cloud-Clone
Supplier Number:  SEF557MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    UCP1
  • Size:
    1 Kit
  • Sample Type:
    Tissue homogenates, cell lysates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Uncoupling Protein 1, Mitochondrial (UCP1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.062 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEF557MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse UCP1 (Uncoupling Protein 1 Mitochondrial). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.062 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

UCP1 is activated in the brown fat cell by fatty acids and inhibited by nucleotides. Sympathetic nervous system terminals release Norepinephrine onto a Beta-3 adrenergic receptor on the plasma membrane. This activates adenylyl cyclase, which catalyses the conversion of ATP to cyclic AMP. cAMP activates protein kinase A, causing its active C subunits to be freed from its regulatory R subunits. Active protein kinase A, in turn, phosphorylates triacylglycerol lipase, thereby activating it. The lipase converts triacylglycerols into free fatty acids, which activate UCP1, overriding the inhibition caused by purine nucleodides. At the termination of thermogenesis, the mitochondria oxidize away the residual fatty acids, UCP1 inactivates and the cell resumes its normal energy-conserving mode.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.