Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse TNFb ELISA Kit
Catalog # MSPP-SEA134MU
CAS Number:  
undefined
Mouse TNFb ELISA Kit
Catalog # MSPP-SEA134MU
Supplier Number:  SEA134MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    TNFb
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Tumor Necrosis Factor Beta (TNFb) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    7.8 - 500 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    2.9 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA134MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse TNFb (Tumor Necrosis Factor Beta). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 2.9 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

TNF-beta is a potent mediator of inflammatory and immune responses.TNF-beta is produced by activated T and B lymphocytes, and has similar activities to TNF-alpha. Like TNF-alpha, TNF-beta is involved in the regulation of various biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, coagulation, and neurotransmission. TNF-beta is secreted as a soluble polypeptide, but can form heterotrimers with lymphotoxin-beta, which effectively anchors the TNF-beta to the cell surface. TNF-beta is cytotoxic to a wide range of tumor cells. LTA is highly inducible, secreted, and exists as homotrimeric molecule. LTA forms heterotrimers with lymphotoxin-beta which anchors lymphotoxin-alpha to the cell surface.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.