Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse SNCa ELISA Kit
Mouse SNCa ELISA Kit
Catalog # MSPP-SEB222MU
CAS Number:  
undefined
Mouse SNCa ELISA Kit
Catalog # MSPP-SEB222MU
Supplier Number:  SEB222MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SNCa
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Synuclein Alpha (SNCa) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    78 - 5000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    30 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB222MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse SNCa (Synuclein Alpha). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 30 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Alpha-synuclein also known as SNCA is a protein .An alpha-synuclein fragment, known as the non-Abeta component (NAC) of Alzheimer's disease amyloid, originally found in an amyloid-enriched fraction, is shown to be a fragment of its precursor protein, NACP, by cloning of the full-length cDNA. Later, NACP turned out to be a human homologue of Torpedo synuclein. Therefore, NACP is also referred to as human alpha-synuclein.

Alpha-synuclein is specifically upregulated in a discrete population of presynaptic terminals of the brain during a period of acquisition-related synaptic rearrangement. It has been shown that alpha-synuclein significantly interacts with tubulin, and that alpha-synuclein may have an activity as potential microtubule-associated protein like tau.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.