Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse SHBG ELISA Kit
Mouse SHBG ELISA Kit
Catalog # MSPP-SEA396MU
CAS Number:  
undefined
Mouse SHBG ELISA Kit
Catalog # MSPP-SEA396MU
Supplier Number:  SEA396MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SHBG
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Sex Hormone Binding Globulin (SHBG) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    31.2 - 2000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    13.1 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA396MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse SHBG (Sex Hormone Binding Globulin). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

SHBG is a glycoprotein that binds to sex hormones, specifically testosterone and estradiol. Other steroid hormones such as progesterone, cortisol, and other corticosteroids are bound by transcortin. These sex hormones circulate in the bloodstream, bound mostly to SHBG and to some degree bound to serum albumin. Only a small fraction is unbound, or 'free,' and thus biologically active and able to enter a cell and activate its receptor. The SHBG inhibits the function of these hormones. Thus bioavailability of sex hormones is influenced by the level of SHBG. SHBG is produced by the liver cells and is released into the bloodstream. Other sites that produce SHBG are the brain, uterus, and placenta and vagina. In addition SHBG is produced by the testes; testes-produced SHBG is also called androgen-binding protein.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.