Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse S100A9 ELISA Kit
Mouse S100A9 ELISA Kit
Catalog # MSPP-SEB793MU
CAS Number:  
undefined
Mouse S100A9 ELISA Kit
Catalog # MSPP-SEB793MU
Supplier Number:  SEB793MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    S100A9
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between S100 Calcium Binding Protein A9 (S100A9) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.1 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB793MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse S100A9 (S100 Calcium Binding Protein A9). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

S100 calcium binding protein A9 has been implicated in the abnormal differentiation of myeloid cells in the stroma of cancer. This contributes to creating an overall immunosuppressive microenvironemnt that may contribute to the inability of a protective or therapeutic cellular immune response to be generated by the tumor-bearing host. Outside of malignancy, S100A9 in association with its dimerization partner, S100A8 (MRP8 or calgranulin A) signals for lymphocyte recruitment in sites of inflammation. S100A9/A8 (synonyma: Calgranulin A/B; Calprotectin) are also regarded as marker proteins for a number of inflammatory diseases in humans, especially in rheumatoid arthritis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.