Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse REG3b ELISA Kit
Mouse REG3b ELISA Kit
Catalog # MSPP-SEK541MU
CAS Number:  
undefined
Mouse REG3b ELISA Kit
Catalog # MSPP-SEK541MU
Supplier Number:  SEK541MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    REG3b
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Regenerating Islet Derived Protein 3 Beta (REG3b) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    9.4 - 600 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    3.3 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEK541MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse REG3b (Regenerating Islet Derived Protein 3 Beta). The assay range is from 9.4 to 600 pg/ml (Sandwich kit) with a sensitivity of 3.3 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

The Reg3 protein family, including the human member designated pancreatitis-associated protein (PAP), consists of secreted proteins that contain a C-type lectin domain involved in carbohydrate binding. They are expressed by intestinal epithelial cells. Colonization of germ-free mice and intestinal infection with pathogens increase the expression of Reg3g and Reg3b in the murine ileum. Reg3g is directly bactericidal for gram-positive bacteria, but the exact role of Reg3b in bacterial infections is unknown. Protein expression of Reg3b and Reg3g was determined in intestinal mucosal scrapings of infected and noninfected mice. Mucosal Reg3b binds to both bacterial pathogens and may interfere with their mode of action. Reg3b plays a protective role against intestinal translocation of the gram-negative bacterium S. enteritidis in mice but not against the gram-positive bacterium L. monocytogenes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.