Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse PALB ELISA Kit
Mouse PALB ELISA Kit
Catalog # MSPP-SEA726MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse PALB ELISA Kit
Catalog # MSPP-SEA726MU
Supplier:  Cloud-Clone
Supplier Number:  SEA726MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    PALB
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, cerebrospinal fluid, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Prealbumin (PALB) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.071 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA726MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse PALB (Prealbumin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.071 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

TTR was originally called prealbumin because it ran faster than albumins on electrophoresis gels.Prealbumin is produced by the choroid plexus, by pancreatic islet cells in the embryonic yolk sac, and by enterochromaffin cells in the gastrointestinal mucosa, but the liver is quantitatively the most important source.9 Liver production is maintained until late in liver disease. Hydration status does not affect prealbumin levels. A negative acute phase reactant, the prealbumin level will transiently decrease in the presence of inflammation and in the immediate postsurgical period. Serum levels also decline in patients with conditions associated with protein malnutrition, such as malignancy, cirrhosis, protein-losing enteropathy, and zinc deficiency.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.