- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Mouse
- Target protein:OC
- Size:1 Kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Osteocalcin (OC) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:31.2 - 2000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:11.3 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA471MU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Mouse OC (Osteocalcin). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 11.3 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Osteocalcin (OC) is the most abundant noncollagenous protein in mature bone where it constitutes 1 to 2% of the total protein. Synthesized by osteoblasts, it is incorporated into the bone matrix. Osteocalcin is a 49-residue protein with three gamma-carboxyglutamic acid residues, at positions 17, 21 and 24. These three residues confer on it a very strong ability to bind to hydroxyapatite. Osteocalcin is a noncollagenous protein found in bone and dentin. It is secreted by osteoblasts and thought to play a role in mineralization and calcium ion homeostasis. It has been stipulated that osteocalcin may also function as a negative regulator of bone formation, although its exact role is unknown.