- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Mouse
- Target protein:NFkB
- Size:1 Kit
- Sample Type:Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Nuclear Factor Kappa B (NFkB) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.057 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEB824MU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Mouse NFkB (Nuclear Factor Kappa B). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.057 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Nuclear factor NF-kappa-B p105 subunit isa 105 kDa protein which can undergo cotranslational processing by the 26S proteasome to produce a 50 kDa protein. The 105 kDa protein is a Rel protein-specific transcription inhibitor and the 50 kDa protein is a DNA binding subunit of the NF-kappaB (NF-κB) protein complex.
NF-κB is a transcription factor that is activated by various intra- and extra-cellular stimuli such as cytokines, oxidant-free radicals, ultraviolet irradiation, and bacterial or viral products. Activated NF-κB translocates into the nucleus and stimulates the expression of genes involved in a wide variety of biological functions, over 200 known genes are targets of NF-κB in various cell types, under specific conditions.