Order Entry
United States
ContactUsLinkComponent
General Species ATP ELISA Kit
General Species ATP ELISA Kit
Catalog # MSPP-CEA349GE
CAS Number:  
undefined
General Species ATP ELISA Kit
Catalog # MSPP-CEA349GE
Supplier Number:  CEA349GE
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    ATP
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Adenosine Triphosphate (ATP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    12.35 - 1000 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    4.33 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA349GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species ATP (Adenosine Triphosphate). The assay range is from 12.35 to 1,000 ng/ml (Competitive kit) with a sensitivity of 4.33 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Adenosine-5'-triphosphate (ATP) is a multifunctional nucleotide that plays an important role in cell biology as a coenzyme, that is, the 'molecular unit of currency' of intracellular energy transfer. ATP transports chemical energy within cells for metabolism. It is an energy source produced during photosynthesis and cellular respiration and consumed by many enzymes and a multitude of cellular processes, including biosynthetic reactions, motility, and cell division. ATP is made from adenosine diphosphate (ADP) or adenosine monophosphate (AMP) and its use in metabolism converts it back into these precursors. ATP is therefore continuously recycled in organisms, with the human body turning over its own weight in ATP each day. ATP was discovered in 1929 by Karl Lohmann, but its correct structure was not determined until some years later. It was proposed to be the main energy-transfer molecule in the cell by Fritz Albert Lipmann in 1941.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.