Order Entry
United States
ContactUsLinkComponent
General Species ADMA ELISA Kit
General Species ADMA ELISA Kit
Catalog # MSPP-CEB301GE
CAS Number:  
undefined
General Species ADMA ELISA Kit
Catalog # MSPP-CEB301GE
Supplier Number:  CEB301GE
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    ADMA
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Asymmetrical Dimethylarginine (ADMA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    12.35 - 1000 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    4.99 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEB301GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species ADMA (Asymmetrical Dimethylarginine). The assay range is from 12.35 to 1,000 ng/ml (Competitive kit) with a sensitivity of 4.99 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Asymmetric dimethylarginine (ADMA) is a naturally occurring chemical found in blood plasma. It is a metabolic by-product of continual protein modification processes in the cytoplasm of all human cells. It is closely related to L-arginine, a conditionally-essential amino acid. ADMA interferes with L-arginine in the production of nitric oxide, a key chemical involved in normal endothelial function and, by extension, cardiovascular health. Nitric oxide (NO) is synthesized in endothelial cells by the enzyme endothelial nitric oxide synthase. The production rate of NO is a key factor for endothelial and cardiovascular functions. The substrate of endothelial nitric oxide synthase is Arg, which is converted into the products NO and citrulline. The enzyme is inhibited by the endogenous Arg metabolite asymmetric dimethylarginine (ADMA).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.