Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine LZM ELISA Kit
Bovine LZM ELISA Kit
Catalog # MSPP-SEB193BO
CAS Number:  
undefined
Bovine LZM ELISA Kit
Catalog # MSPP-SEB193BO
Supplier Number:  SEB193BO
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Bovine
  • Target protein:
    LZM
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Lysozyme (LZM) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.131 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB193BO
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Bovine LZM (Lysozyme). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.131 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Lysozyme, also known as muramidase or N-acetylmuramide glycanhydrolase, are a family of enzymes (EC 3.2.1.17) which damage bacterial cell walls by catalyzing hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in a peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrins. Lysozyme is abundant in a number of secretions, such as tears, saliva, human milk and mucus. It is also present in cytoplasmic granules of the polymorphonuclear neutrophils (PMN). Large amounts of lysozyme can be found in egg white. C-type lysozymes are closely related to alpha-lactalbumin in sequence and structure making them part of the same family.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.