- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Bovine
- Target protein:IL8
- Size:1 Kit
- Sample Type:Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Interleukin 8 (IL8) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:15.6 - 1000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:6.2 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA080BO
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Bovine IL8 (Interleukin 8). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interleukin 8 (IL-8), a member of the neutrophil-specific CXC subfamily of chemokines, is a potent neutrophil chemotactic and activating factor. It is a primary inflammatory cytokine produced by many cells in response to proinflammatory stimuli such as IL-1, TNF, LPS and viruses. Its function is, in part, to attract neutrophils to the site of inflammation and to activate them. The IL-8 cDNA sequence predicts a protein of 99 amino acids. Removal of a 22-residue signal peptide generates a mature protein of 77 amino acids (~8 kDa). Further proteolysis of the N-terminal end leads to a variant form with 72 amino acids, full activation of IL-8 may require cleavage to the 72 amino acid form. IL-8 can form non-covalent dimers in solution, especially at high concentrations, but dimerization is not necessary for biological activity.