Order Entry
Puerto Rico
ContactUsLinkComponent
Rat PINP ELISA Kit
Rat PINP ELISA Kit
Catalog # MSPP-CEA957RA
Supplier:  Cloud-Clone
CAS Number:  
undefined
Rat PINP ELISA Kit
Catalog # MSPP-CEA957RA
Supplier:  Cloud-Clone
Supplier Number:  CEA957RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    PINP
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Procollagen I N-Terminal Propeptide (PINP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    246.9 - 20000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    95.3 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA957RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat PINP (Procollagen I N to Terminal Propeptide). The assay range is from 246.9 to 20,000 pg/ml (Competitive kit) with a sensitivity of 95.3 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

The PINP molecule is similar to PIIINP consisting of three distinct structural domains: Col 1 is on the aminoterminal side of the molecule, while Col 2 and Col 3 are situated on the middle of the helically structured molecule (Kühn et al., 1982). The PINP molecule has a molecular mass of 35 000 and is cleared by scavenger receptors in liver endothelial cells (Melkko et al., 1994). PINP often occurs in circulation in two forms of different molecular sizes. One is identical to the trimeric authentic antigen (intact PINP) whereas the other consists of smaller forms of PINP, resembling a single domain of the proα1 (I) chain of PINP and is probably a degradation product of type I procollagen or I pN-collagen. Thus, an assay of intact PINP rather than total PINP appears to be more sensitive in detecting changes in the rate of type I collagen synthesis (Melkko et al., 1996, Risteli & Risteli 1999).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.