Order Entry
Puerto Rico
ContactUsLinkComponent
Rat NT-ProBNP ELISA Kit
Rat NT-ProBNP ELISA Kit
Catalog # MSPP-CEA485RA
CAS Number:  
undefined
Rat NT-ProBNP ELISA Kit
Catalog # MSPP-CEA485RA
Supplier Number:  CEA485RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    NT-ProBNP
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    123.5 - 10000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    51.1 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA485RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat NT to ProBNP (N to Terminal Pro to Brain Natriuretic Peptide). The assay range is from 123.5 to 10,000 pg/ml (Competitive kit) with a sensitivity of 51.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Brain-type Natriuretic Peptide (BNP) is a nonglycosylated peptide that is produced predominantly by ventricular myocytes and belongs to the natriuretic peptide family. Plasma NT-proBNP is a marker for congestive heart failure, while mature BNP (aa 103 - 134) promotes vasodilation and fluid and sodium excretion.BNP is co-secreted along with a 76 amino acid N-terminal fragment (NT-proBNP) which is biologically inactive. BNP binds to and activates the atrial natriuretic factor receptors NPRA, and to a lesser extent NPRB, in a fashion similar to atrial natriuretic peptide (ANP) but with 10-fold lower affinity. The biological half-life of BNP, however, is twice as long as that of ANP, and that of NT-proBNP is even longer, making these peptides better targets than ANP for diagnostic blood testing.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.