Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse TNFa ELISA Kit
Mouse TNFa ELISA Kit
Catalog # MSPP-SCA133MU
CAS Number:  
undefined
Mouse TNFa ELISA Kit
Catalog # MSPP-SCA133MU
Supplier Number:  SCA133MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    TNFa
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha (TNFa) and analogues was observed
  • Detection Method:
    Chemiluminescence
  • Time to Results:
    2 h, 40 min
  • Shelf Life:
    12 Months
  • Detection Range:
    1.37 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.53 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SCA133MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse TNFa (Tumor Necrosis Factor Alpha). The assay range is from 1.37 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 0.53 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

TNFa  is synthesized as a 26 kDa, type II transmembrane protein that is 233 amino acids in length. It contains a 30 amino acid (aa) cytoplasmic domain, a 26 aa transmembrane segment, and a 177 aa extracellular region. TNFa is assembled intracellularly to form a transmembrane, non-covalently-linked homotrimeric protein. The 157 aa residue soluble form of TNFa (sTNF-αis released from the C-terminus of the transmembrane protein through the activity of TNFa-converting enzyme (TACE), a membrane -bound disintegrin metalloproteinase. Rat cells known to express TNF-αinclude B cells, colonic columnar epithelial cells, NK and CD3 CD56 hepatic natural T cells, macrophages, monocytes and monocyte-derived dendritic cells, CD4 and CD8 T cells, mast cells, neutrophils, keratinocytes, plasma cells, and adipocytes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.