- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Mouse
- Target protein:TNFa
- Size:1 Kit
- Sample Type:Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha (TNFa) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:15.6 - 1000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:5.7 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA133MU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Mouse TNFa (Tumor Necrosis Factor Alpha). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
TNFa is synthesized as a 26 kDa, type II transmembrane protein that is 233 amino acids in length. It contains a 30 amino acid (aa) cytoplasmic domain, a 26 aa transmembrane segment, and a 177 aa extracellular region. TNFa is assembled intracellularly to form a transmembrane, non-covalently-linked homotrimeric protein. The 157 aa residue soluble form of TNFa (sTNF-αis released from the C-terminus of the transmembrane protein through the activity of TNFa-converting enzyme (TACE), a membrane -bound disintegrin metalloproteinase. Rat cells known to express TNF-αinclude B cells, colonic columnar epithelial cells, NK and CD3 CD56 hepatic natural T cells, macrophages, monocytes and monocyte-derived dendritic cells, CD4 and CD8 T cells, mast cells, neutrophils, keratinocytes, plasma cells, and adipocytes.