Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse INHbE ELISA Kit
  MSPP-SEA048MU
 :  
undefined
Mouse INHbE ELISA Kit
  MSPP-SEA048MU
 :  SEA048MU
 :  

 

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    INHbE
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Inhibin Beta E (INHbE) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.9 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA048MU
  • No. of tests:
    96 wells

 

 

This assay has high sensitivity and excellent specificity for detecting Mouse INHbE (Inhibin Beta E). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 5.9 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

INHBE is a member of the activin beta family that plays a role in pancreatic exocrine cell growth and proliferation.Hashimoto et al., cloned INHBE, which they called activin beta-E subunit. The deduced 350-amino acid protein contains an N-terminal hydrophobic signal peptide and a potential N-glycosylation site. The proposed cleavage site at amino acids 232 to 236 would yield a mature 114-amino acid protein., INHBE shares 97 and 96% similarity with the mouse and rat activin beta-E orthologs, respectively. The deduced mature human beta-E subunit shares amino acid identity of 44, 48 and 64% with activin beta-A, activin beta-B, and activin beta-C, respectively.

 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.