Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse IL6 ELISA Kit
Mouse IL6 ELISA Kit
Catalog # MSPP-SEA079MU
CAS Number:  
undefined
Mouse IL6 ELISA Kit
Catalog # MSPP-SEA079MU
Supplier Number:  SEA079MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL6
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 6 (IL6) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    7.8 - 500 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    3.0 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA079MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse IL6 (Interleukin 6). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 3.0 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

IL-6 is a multifunctional protein produced by lymphoid and non-lymphoid cells, and by normal and transformed cells, including T cells, monocyte/macrophages, fibroblasts, hepatocytes, vascular endothelial cells, cardiac myxomas, bladder cell carcinomas, myelomas, astrogliomas and glioblastomas. The production of IL-6 in these various cells is regulated, either positively or negatively, by a variety of signals including mitogens, antigenic stimulation, lipopolysaccharides, IL-1, TNF, PDGF and viruses. On the basis of its various activities, IL-6 has also been called interferon-β2 (IFN-β2), 26 kDa protein, B-cell stimulatory factor-2 (BSF-2), hybridoma/plasmacytoma growth factor, hepatocyte stimulating factor, cytotoxic T-cell differentiation factor, and macrophage-granulocyte inducing factor 2A (MGI-2A).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.