To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:IgG
- Size:1 Kit
- Environmentally Preferable:
- Sample Type:Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Immunoglobulin G (IgG) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:6.25 - 400 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:2.87 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA544HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human IgG (Immunoglobulin G). The assay range is from 6.25 to 400 ng/ml (Sandwich kit) with a sensitivity of 2.87 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Immunoglobulin G (IgG) is a monomeric immunoglobulin, built of two heavy chains γ and two light chains. Each IgG has two antigen binding sites. It is the most abundant immunoglobulin and is approximately equally distributed in blood and in tissue liquids, constituting 75% of serum immunoglobulins in humans. IgG molecules are synthesised and secreted by plasma B cells. IgG antibodies are predominately involved in the secondary antibody response, which occurs approximately one month following antigen recognition, thus the presence of specific IgG generally corresponds to maturation of the antibody response. This is the only isotype that can pass through the human placenta, thereby providing protection to the fetus in utero. Along with IgA secreted in the breast milk, residual IgG absorbed through the placenta provides the neonate with humoral immunity before its own immune system develops.