- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:CAMP
- Size:1 Kit
- Sample Type:Serum, Plasma and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Cathelicidin Antimicrobial Peptide (CAMP) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:0.78 - 50 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.27 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEC419HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human CAMP (Cathelicidin Antimicrobial Peptide). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.27 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Cathelicidin antimicrobial peptide is a family of polypeptides found in lysosomes in polymorphonuclear leukocytes (PMNs). Members of the cathelicidin family of antimicrobial polypeptides are characterized by a highly conserved region (cathelin domain) and a highly variable cathelicidin peptide domain. Cathelicidin peptides have been isolated from many different species of mammals. Cathelicidins were originally found in neutrophils but have since been found in many other cells including epithelial cells and macrophages activated by bacteria, viruses, fungi, or the hormone 1,25-D. The cathelicidin family shares primary sequence homology with the cathepsin family of cysteine proteinase inhibitors, although amino acid residues thought to be important in such protease inhibition are usually lacking.