Order Entry
Puerto Rico
ContactUsLinkComponent
Human Ab1-42 ELISA Kit
Human Ab1-42 ELISA Kit
Catalog # MSPP-CEA946HU
CAS Number:  
undefined
Human Ab1-42 ELISA Kit
Catalog # MSPP-CEA946HU
Supplier Number:  CEA946HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    Ab1-42
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Amyloid Beta Peptide 1-42 (Ab1-42) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.70 - 300 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    1.44 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA946HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human Ab1 to 42 (Amyloid Beta Peptide 1 to 42). The assay range is from 3.70 to 300 pg/ml (Competitive kit) with a sensitivity of 1.44 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

One of the hallmarks of Alzheimer's disease is the self-aggregation of the amyloid β peptide (Aβ) in extracellular amyloid fibrils. Aβ is a peptide composed of 40 to 42 (43) amino acids, and is said to be cleaved out of the precursor protein APP by the action of β- or γ-secretase.In addition, the presence of numerous variant A β molecules has been demonstrated in the culture fluid of mouse neuroblastoma cells transfected with cDNA coding human amyloid precursor protein (APP). Among the different forms of Aβ, the 42-residue fragment (Aβ1-42) readily self-associates and forms nucleation centers from where fibrils can quickly grow. The strong tendency of Aβ1-42 to aggregate is one of the reasons for the scarcity of data on its fibril formation process. Aβ1-40 is the most common form secreted from cultured cells and found in cerebro-spinal fluid (CSF). Aβ1-42 is the major component of senile plaques and considered as the most crucial factor in AD pathogenesis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.