Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse MPO ELISA Kit
Mouse MPO ELISA Kit
Catalog # MSPP-SEA601MU
CAS Number:  
undefined
Mouse MPO ELISA Kit
Catalog # MSPP-SEA601MU
Supplier Number:  SEA601MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MPO
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Myeloperoxidase (MPO) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    78 - 5000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    31 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA601MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse MPO (Myeloperoxidase). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 31 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Myeloperoxidase (MPO) is a hemoprotein that is abundantly expressed in polymorphonuclear leukocytes (neutrophils) and secreted during their activation. The presence of a peroxidase in the cytoplasmic granules of leukocytes was suggested at the beginning of 20th century but it was the early 1940s that it was purified for the first time. Native MPO is a covalently bound tetrameric complex of two glycosylated alpha chains (MW 59 – 64 kDa) and two unglycosylated beta chains (MW 14 kDa) with total MW about 150 kDa and theoretical pI 9.2. MPO plays an important role in neutrophil microbicidal action through catalyzing chloride ion oxidation to hypochlorous acid, which is a potent antimicrobial agent.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.