Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse MOTS-c ELISA Kit
Catalog # MSPP-CEX132MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse MOTS-c ELISA Kit
Catalog # MSPP-CEX132MU
Supplier:  Cloud-Clone
Supplier Number:  CEX132MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MOTS-c
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Mitochondrial Open Reading Frame Of The 12S rRNA-c (MOTS-c) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    246.9 - 20000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    98.7 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEX132MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse MOTS-c (Mitochondrial Open Reading Frame Of The 12S rRNA-c). The assay range is from 246.9 to 20000 pg/ml (Competitive kit) with a sensitivity of 98.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Mitochondria are known to be functional organelles, but their role as a signaling unit is increasingly being appreciated. The identification of a short open reading frame (sORF) in the mitochondrial DNA (mtDNA) that encodes a signaling peptide, humanin, suggests the possible existence of additional sORFs in the mtDNA. A sORF within the mitochondrial 12S rRNA encoding a 16-amino-acid peptide named MOTS-c that regulates insulin sensitivity and metabolic homeostasis. Its primary target organ appears to be the skeletal muscle, and its cellular actions inhibit the folate cycle and its tethered de novo purine biosynthesis, leading to AMPK activation. MOTS-c treatment in mice prevented age-dependent and high-fat-diet-induced insulin resistance, as well as diet-induced obesity. These results suggest that mitochondria may actively regulate metabolic homeostasis at the cellular and organismal level via peptides encoded within their genome.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.