Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse MMP9 ELISA Kit
Mouse MMP9 ELISA Kit
Catalog # MSPP-SEA553MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse MMP9 ELISA Kit
Catalog # MSPP-SEA553MU
Supplier:  Cloud-Clone
Supplier Number:  SEA553MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MMP9
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Matrix Metalloproteinase 9 (MMP9) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.059 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA553MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse MMP9 (Matrix Metalloproteinase 9). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

MMP-9 (also known as gelatinase B) is secreted as a 92 kDa zymogen. Cleavage of pro-MMP-9 at or near residue 87 results in the active enzyme with a mass of approximately 82 kDa. MMP-9 has three fibronectin type II domains, a hemopexin-like domain and a proline-rich type V collagen-homologous domain. Pro-MMP-9 can be activated by MMP-3 or by certain bacterial proteinases. MMP-9 is inhibited byα2-macroglobulin or by TIMP-1, which binds to pro-MMP-9 as well as to active MMP-9.Pro-MMP-9 is secreted by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts, chondrocytes, skeletal muscle satellite cells, endothelial cells, and various tumor cells. Pro-MMP-9 expression is upregulated by TGF-β1, IL-1β, TGF-α, PDGF-AB,

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.