Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse LTA4H ELISA Kit
Mouse LTA4H ELISA Kit
Catalog # MSPP-SED236MU
CAS Number:  
undefined
Mouse LTA4H ELISA Kit
Catalog # MSPP-SED236MU
Supplier Number:  SED236MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    LTA4H
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Leukotriene A4 Hydrolase (LTA4H) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.065 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SED236MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse LTA4H (Leukotriene A4 Hydrolase). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.065 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Minami et al., (1987) reported the full-length cDNA and complete primary structure of human LTA4 hydrolase. This was the first report of the molecular cloning of an enzyme involved in the biosynthesis of eicosanoids. Funk et al., (1987) isolated a cDNA clone corresponding to leukotriene A4 hydrolase from a human lung lambda-gt11 expression library by immunoscreening with a polyclonal antiserum. Several additional clones from human lung and placenta cDNA lambda-gt11 libraries were obtained by plaque hybridization with the (32)P-labeled lung cDNA clone. One of the clones had an insert of 1910 basepairs that contained a complete protein-coding region. From the deduced primary structure, leukotriene A4 hydrolase is a 610-amino acid protein with a calculated molecular weight of 69.140.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.