Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse LOXL2 ELISA Kit
Mouse LOXL2 ELISA Kit
Catalog # MSPP-SEF552MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse LOXL2 ELISA Kit
Catalog # MSPP-SEF552MU
Supplier:  Cloud-Clone
Supplier Number:  SEF552MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    LOXL2
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Lysyl Oxidase Like Protein 2 (LOXL2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.063 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEF552MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse LOXL2 (Lysyl Oxidase Like Protein 2). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.063 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Lysyl oxidase homolog 2 is encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.Expressed in many tissues. Highest expression in reproductive tissues, placenta, uterus and prostate.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.