Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse IL17 ELISA Kit
Mouse IL17 ELISA Kit
Catalog # MSPP-SEA063MU
CAS Number:  
undefined
Mouse IL17 ELISA Kit
Catalog # MSPP-SEA063MU
Supplier Number:  SEA063MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL17
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 17 (IL17) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.3 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA063MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse IL17 (Interleukin 17). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.3 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Interleukin 17 (IL-17; also known as IL-17A) is a 30 to 35 kDa variably glycosylated homodimeric protein that belongs to a unique family of cysteine-knot related proteins. Its sequence was originally isolated from an activated rodent hybridoma and termed CTLA-8. It is synthesized as a 155 amino acid (aa) precursor that contains a 23 aa signal sequence and a 15 kDa, 132 aa mature segment. Although there are two intrachain disulfide bonds that create a ring reminiscent of those found in cysteine-knot proteins, the actual closed knot structure does not appear to form. IL-17 has one potential N-linked glycosylation site.IL-17, a T cell-derived proinf lammatory cytokine, is produced by TCRα/β+CD4-CD8- thymocytes, as well as by activated CD4+ and CD4+CD45RO+ memory T cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.