Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse GDF15 ELISA Kit
Mouse GDF15 ELISA Kit
Catalog # MSPP-SEC034MU
CAS Number:  
undefined
Mouse GDF15 ELISA Kit
Catalog # MSPP-SEC034MU
Supplier Number:  SEC034MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    GDF15
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Growth Differentiation Factor 15 (GDF15) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.056 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEC034MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse GDF15 (Growth Differentiation Factor 15). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

GDF15 is a protein belonging to the transforming growth factor beta superfamily that has a role in regulating inflammatory and apoptotic pathways in injured tissues and during disease processes.GDF15 is also known as TGF-PL, MIC-1, PDF, PLAB, and PTGFB.GDF15 mRNA is most abundant in the liver, with lower levels seen in some other tissues. Its expression in liver can be significantly up-regulated in during injury of organs such as liver, kidney, heart and lung. Northern blot analysis showed that MIC1 is not expressed in undifferentiated cells but is progressively upregulated upon differentiation with retinoic acid followed by phorbol ester or cytokine treatment. Exposure of macrophages to MIC1 or TGFB1, followed by stimulation with lipopolysaccharide, suppressed the production of TNFA.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.