Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse FLG ELISA Kit
Catalog # MSPP-SEJ103MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse FLG ELISA Kit
Catalog # MSPP-SEJ103MU
Supplier:  Cloud-Clone
Supplier Number:  SEJ103MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    FLG
  • Size:
    1 kit
  • Sample Type:
    Tissue homogenates, cell lysates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Filaggrin (FLG) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.114 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEJ103MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse FLG (Filaggrin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.114 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Filaggrinis an intermediate filament-associated protein that aggregates keratin intermediate filaments in mammalian epidermis. It is initially synthesized as a polyprotein precursor, profilaggrin (consisting of multiple filaggrin units of 324 aa each), which is localized in keratohyalin granules, and is subsequently proteolytically processed into individual functional filaggrin molecules. Mutations in this gene are associated with ichthyosis vulgaris. This structure is similar to that of the mouse; however, the human filaggrin repeat is much longer (972 basepairs; 324 amino acids) and shows little sequence homology to the mouse protein. Amino acid sequences encoding the amino and carboxyl termini were more conserved, as were the 5-prime and 3-prime DNA sequences flanking the coding portions of the gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.