Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse FGL2 ELISA Kit
Mouse FGL2 ELISA Kit
Catalog # MSPP-SEA512MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse FGL2 ELISA Kit
Catalog # MSPP-SEA512MU
Supplier:  Cloud-Clone
Supplier Number:  SEA512MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    FGL2
  • Size:
    1 kit
  • Sample Type:
    Plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Fibrinogen Like Protein 2 (FGL2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.135 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA512MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse FGL2 (Fibrinogen Like Protein 2). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.135 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

FGL2 is a 439 amino acid secreted protein that is similar to the β- and γ-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. This protein may play a role in physiologic functions at mucosal sites.Fgl2 is a protein that exhibits pleiotropic effects within the body and is an important immune regulator of both innate and adaptive responses. The protein exists as both a type II transmembrane protein found on the surface of macrophages and endothelial cells and can be constitutively secreted by both CD4+ and CD8+ T cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.