Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse DEFb2 ELISA Kit
Mouse DEFb2 ELISA Kit
Catalog # MSPP-SEA072MU
CAS Number:  
undefined
Mouse DEFb2 ELISA Kit
Catalog # MSPP-SEA072MU
Supplier Number:  SEA072MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    DEFb2
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Defensin Beta 2 (DEFb2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.0 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA072MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse DEFb2 (Defensin Beta 2). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.0 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Beta-defensin 2 is a protein encoded by the DEFB4 gene.Defensins are 2-6 kDa, cationic, microbicidal peptides active against many Gram-negative and Gram-positive bacteria, fungi, and enveloped viruses, containing three pairs of intramolecular disulfide bonds. On the basis of their size and pattern of disulfide bonding, mammalian defensins are classified into alpha, beta and theta categories. Every mammalian species explored thus far has beta-defensins. In cows, as many as 13 beta-defensins exist in neutrophils. However, in other species, beta-defensins are more often produced by epithelial cells lining various organs. Human, rabbit and guinea-pig beta-defensins, as well as human beta-defensin-2 (hBD2), induce the activation and degranulation of mast cells, resulting in the release of histamine and prostaglandin D2.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.