Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse CXCL2 ELISA Kit
Mouse CXCL2 ELISA Kit
Catalog # MSPP-SEB603MU
CAS Number:  
undefined
Mouse CXCL2 ELISA Kit
Catalog # MSPP-SEB603MU
Supplier Number:  SEB603MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    CXCL2
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Chemokine (C-X-C Motif) Ligand 2 (CXCL2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB603MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse CXCL2 (Chemokine (C-X-C Motif) Ligand 2). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Growth-regulated oncogene (GRO)beta, which belongs to the CXC chemokine family, is a potent chemoattractant for neutrophils.Chemokine (C-X-C motif) ligand 2 (CXCL2) is a small cytokine belonging to the CXC chemokine family that is also called macrophage inflammatory protein 2-alpha (MIP2-alpha), Growth-regulated protein beta (Gro-beta) and Gro oncogene-2 (Gro-2). CXCL2 is 90% identical in amino acid sequence as a related chemokine, CXCL1. This chemokine is secreted by monocytes and macrophages and is chemotactic for polymorphonuclear leukocytes and hematopoietic stem cells. The gene for CXCL2 is located on human chromosome 4 in a cluster of other CXC chemokines.CXCL2 mobilizes cells by interacting with a cell surface chemokine receptor called CXCR2.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.