Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse CXCL14 ELISA Kit

Mouse CXCL14 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEB607MU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEB607MU
Mouse CXCL14 ELISA Kit
Catalog # MSPP-SEB607MU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Mouse CXCL14 ELISA Kit
Mouse CXCL14 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$684.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Mouse CXCL14 (Chemokine (C-X-C Motif) Ligand 14). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Chemokine (C-X-C motif) ligand 14 (CXCL14) is a small cytokine belonging to the CXC chemokine family that is also known as BRAK (for breast and kidney-expressed chemokine).Mature CXCL14 has many of the conserved features of the CXC chemokine subfamily but has some differences too, such as a shorter N-terminus and five extra amino acids in the region between its third and fourth cysteines. CXCL14 is constitutively expressed at high levels in many normal tissues, where its cellular source is thought to be fibroblasts.However, it is reduced or absent from most cancer cells.This chemokine is chemotactic for monocytes and can activate these cells in the presence of an inflammatory mediator called prostaglandin-E2 (PGE2).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    0.156 - 10 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEB607MU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.059 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    CXCL14
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Chemokine (C-X-C Motif) Ligand 14 (CXCL14) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.