Mouse CPP ELISA Kit
This assay has high sensitivity and excellent specificity for detecting Mouse CPP (Copeptin). The assay range is from 24.69 to 2000 pg/ml (Competitive kit) with a sensitivity of 9.16 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Copeptin, a stable peptide of the arginine vasopressin precursor, is elevated in hemorrhagic and septic shock.Arginine vasopressin (AVP) levels are increased in hemorrhagic and septic shock. Measurement of AVP levels has limitations due to its short half-life and cumbersome detection method. Copeptin is a more stable peptide derived from the same precursor molecule.C-Terminal Provasopressin (Copeptin) as a Novel and Prognostic Marker in Acute Myocardial Infarction. arginine vasopressin-neurophysin II isa precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin II and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus.
: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.
- Size:1 kit
- No. of tests:96 wells
- Primary antibody reactivity:Mouse
- Detection Range:24.69 - 2000 pg/ml
- Sample Volume:50 µl
- Host:Rabbit
- Time to Results:2 h
- Detection Method:Colorimetric
- Cat. No.:MSPP-CEA365MU
- Regulatory Status:RUO
- Assay duration:Multiple steps
- Sensitivity:9.16 pg/ml
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Assay Type (ELISA with LOV):Competitive
- Target protein:CPP
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Shelf Life:12 Months
- Cross Reactivity:No significant cross-reactivity or interference between Copeptin (CPP) and analogues was observed
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



