Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse CCK ELISA Kit

Mouse CCK ELISA Kit

96 wells, 1 kit
Catalog # MSPP-CEA802MU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  CEA802MU
Mouse CCK ELISA Kit
Catalog # MSPP-CEA802MU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Mouse CCK ELISA Kit
Mouse CCK ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$648.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Mouse CCK (Cholecystokinin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.61 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Cholecystokinin(CCK) plays a key role in facilitating digestion within the small intestine. It is secreted from mucosal epithelial cells in the first segment of the small intestine (duodenum), and stimulates delivery into the small intestine of digestive enzymes from the pancreas and bile from the gallbladder. Cholecystokinin is also produced by neurons in the enteric nervous system, and is widely and abundantly distributed in the brain. Cholecystokinin and gastrin are highly similar peptides. Like gastrin, cholecystokinin is a linear peptide that is synthesized as a preprohormone, then proteolytically cleaved to generate a family of peptides having the same carboxy ends. Full biologic activity is retained in CCK-8 (8 amino acids), but peptides of 33, 38 and 59 amino acids are also produced.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    12.35 - 1000 pg/ml
  • Sample Volume:
    50 µl
  • Host:
    Rabbit
  • Time to Results:
    2 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-CEA802MU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    4.61 pg/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Competitive
  • Target protein:
    CCK
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Cholecystokinin (CCK) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.