Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse bEP ELISA Kit
Mouse bEP ELISA Kit
Catalog # MSPP-CEA806MU
CAS Number:  
undefined
Mouse bEP ELISA Kit
Catalog # MSPP-CEA806MU
Supplier Number:  CEA806MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    bEP
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, cerebrospinal fluid and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Beta-Endorphin (bEP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    12.35 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    4.74 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA806MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse bEP (Beta-Endorphin). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 4.74 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Beta-endorphin is an endogenous opioid peptide neurotransmitter found in the neurons of both the central and peripheral nervous system.Sequence: 'Tyr Gly Gly Phe Met Thr Ser Glu Lys Ser Gln Thr Pro Leu Val Thr Leu Phe Lys Asn Ala Ile Ile Lys Asn Ala Tyr Lys Lys Gly Glu'. β-endorphin is a peptide, 31 amino acids long, resulting from processing of the precursor proopiomelanocortin (POMC). (Note, POMC also gives rise to other peptide hormones, including ACTH, as well α- and γ-MSH, resulting from intracellular processing by internal enzymes known as prohormone convertases.) β-endorphin is found in neurons of the hypothalamus, as well as the pituitary gland.It is an agonist of the opioid receptors, with evidence suggesting it serves as the endogenous ligand of the μ-opioid receptor, the same receptor to which the chemicals extracted from opium, such as morphine and codeine, have their analgesic and addictive effects.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.