Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse AMH ELISA Kit
Catalog # MSPP-CEA228MU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse AMH ELISA Kit
Catalog # MSPP-CEA228MU
Supplier:  Cloud-Clone
Supplier Number:  CEA228MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    AMH
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Anti-Mullerian Hormone (AMH) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    123.5 - 10000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    50.2 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA228MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse AMH (Anti-Mullerian Hormone). The assay range is from 123.5 to 10000 pg/ml (Competitive kit) with a sensitivity of 50.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Anti-Müllerian hormone (AMH) is a dimeric glycoprotein that inhibits the development of the Müllerian ducts in a male embryo. It is named after Johannes Peter Müller. AMH is a protein hormone structurally related to inhibin and activin, and a member of the transforming growth factor-β (TGF-β) family. It is present in fish, reptiles, birds, marsupials, and placental mammals.In mammals AMH is secreted by Sertoli cells of the testes during embryogenesis of the fetal male and prevents the development of the mullerian ducts into the uterus and other mullerian structures. The effect is ipsilateral, that is each testis suppresses Müllerian development only on its own side. In humans this action takes place by 8 weeks gestation. In female embryogenesis the absence of AMH allows for the development of upper vagina, uterus and cervix, and oviducts.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.