Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse ABP1 ELISA Kit

Mouse ABP1 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEA656MU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEA656MU
Mouse ABP1 ELISA Kit
Catalog # MSPP-SEA656MU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Mouse ABP1 ELISA Kit
Mouse ABP1 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$648.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Mouse ABP1 (Amiloride Binding Protein 1). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

On the basis of its primary structure, the amiloride-binding protein (EC 1.4.3.6) is 713 amino acids long, with a 19-amino acid signal peptide. Expressed in cultured cells, the mRNA yields a glycoprotein that binds amiloride and amiloride analogs with affinities similar to the amiloride receptor associated with the apical Na⁺ channel in pig kidney membranes and is immunoprecipitated with monoclonal antibodies raised against pig kidney amiloride-binding protein. Barbry et al., (1990) pointed out that amiloride-sensitive Na⁺ channels are also present in airway epithelia, where they play an important role in fluid secretion. Amiloride inhibits the excessive absorption of Na⁺ and liquid that takes place in airway epithelia of patients with cystic fibrosis, and amiloride aerosol therapy has been tried for the treatment of lung disease in CF.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    31.2 - 2000 pg/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEA656MU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    13.2 pg/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    ABP1
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Amiloride Binding Protein 1 (ABP1) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.