Order Entry
Puerto Rico
ContactUsLinkComponent
Human vWFCP ELISA Kit
Human vWFCP ELISA Kit
Catalog # MSPP-SEA950HU
CAS Number:  
undefined
Human vWFCP ELISA Kit
Catalog # MSPP-SEA950HU
Supplier Number:  SEA950HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    vWFCP
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Von Willebrand Factor Cleaving Protease (vWFCP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.115 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA950HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human vWFCP (Von Willebrand Factor Cleaving Protease). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.115 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

ADAMTS13 (a disintegrin and metalloproteinase with a thrombospondin type 1 motif, member 13)—also known as von Willebrand factor-cleaving protease (VWFCP)—is a zinc-containing metalloprotease enzyme that cleaves von Willebrand factor (vWf), a large protein involved in blood clotting. It is secreted in blood and degrades large vWf multimers, decreasing their activity.Genomically, ADAMTS13 shares many properties with the 19 member ADAMTS family, all of which are characterised by a protease domain (the part that performs the protein hydrolysis), an adjacent disintegrin domain and one or more thrombospondin domains. ADAMTS13 in fact has eight thrombospondin domains. It has no hydrophobic transmembrane domain, and hence it not anchored in the cell membrane.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.