Order Entry
Puerto Rico
ContactUsLinkComponent
Human Vaspin ELISA Kit
Human Vaspin ELISA Kit
Catalog # MSPP-SEA706HU
CAS Number:  
undefined
Human Vaspin ELISA Kit
Catalog # MSPP-SEA706HU
Supplier Number:  SEA706HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    Vaspin
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Visceral Adipose Tissue Derived Serine Protease Inhibitor (Vaspin) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.054 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA706HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human Vaspin (Visceral Adipose Tissue Derived Serine Protease Inhibitor). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Visceral adipose tissue-derived serine protease inhibitor (vaspin) is an interesting novel adipocytokine with insulin-sensitizing effects. Some studies have suggested that vaspin could play an important role in the development of obesity and metabolic disorders. The induction of vaspin mRNA expression could represent a compensatory mechanism associated with obesity, severe insulin resistance and type 2 diabetes mellitus, however it is unclear whether a correlation exists between human vaspin serum levels and markers of insulin sensitivity and glucose or lipid metabolism. Vaspin serum concentrations have been shown to be lower in lean subjects and competitive sportsmen with long-term physical training, but they are increased with weight loss associated with a physical training programme.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.