Human Trx ELISA Kit
This assay has high sensitivity and excellent specificity for detecting Human Trx (Thioredoxin). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.22 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Thioredoxins are proteins that act as antioxidants by facilitating the reduction of other proteins by cysteine thiol-disulfide exchange. Thioredoxins are found in nearly all known organisms and are essential for life in mammals.Thioredoxins are characterized at the level of their amino acid sequence by the presence of two vicinal cysteines in a CXXC motif. These two cysteines are the key to the ability of thioredoxin to reduce other proteins. Thioredoxin proteins also have a characteristic tertiary structure termed the thioredoxin fold.The thioredoxins are kept in the reduced state by the flavoenzyme thioredoxin reductase, in a NADPH-dependent reaction. Thioredoxins act as electron donors to peroxidases and ribonucleotide reductase.
: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.
- Size:1 kit
- No. of tests:96 wells
- Primary antibody reactivity:Human
- Detection Range:3.12 - 200 ng/ml
- Sample Volume:100 µl
- Host:Rabbit
- Time to Results:3 h
- Detection Method:Colorimetric
- Cat. No.:MSPP-SEA702HU
- Regulatory Status:RUO
- Assay duration:Multiple steps
- Sensitivity:1.22 ng/ml
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Assay Type (ELISA with LOV):Sandwich
- Target protein:Trx
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Shelf Life:12 Months
- Cross Reactivity:No significant cross-reactivity or interference between Thioredoxin (Trx) and analogues was observed
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year



