- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:TJP1
- Size:1 kit
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Tight Junction Protein 1 (TJP1) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.059 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEC262HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human TJP1 (Tight Junction Protein 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Tight junction protein ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. The encoded protein may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for this gene. Tight junction (zonula occludens) protein-1 (TJP1), also referred to as ZO-1, is a 200 kDa protein located on a cytoplasmic membrane surface of vertebrate intercellular tight junctions. Willott et al., (1993) isolated a full-length cDNA sequence for human TJP1. Although the function of TJP1 is unknown, the cDNA sequence predicted a multi-domain signaling protein homologous to the product of the 'discs large-1' tumor suppressor gene of Drosophila and several other membrane-associated proteins in mammals.