Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human STAT3 ELISA Kit

Human STAT3 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEB743HU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEB743HU
Human STAT3 ELISA Kit
Catalog # MSPP-SEB743HU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human STAT3 ELISA Kit
Human STAT3 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$700.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Human STAT3 (Signal Transducer And Activator Of Transcription 3). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.062 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

STAT3, a transcription factor for acute phase response genes, acts as an adaptor molecule in signal transduction from the type I interferon receptor. They found that it binds to a conserved sequence in the cytoplasmic tail of the IFNAR1 chain of the receptor and undergoes interferon-dependent tyrosine phosphorylation. The p85 regulatory subunit of phosphatidylinositol 3-kinase, which activates a series of serine kinases, was found to bind to phosphorylated STAT3 and subsequently to undergo tyrosine phosphorylation. The authors concluded that STAT3 acts as an adaptor to couple another signaling pathway to the interferon receptor.In many human cancers and transformed cell lines, STAT3 is persistently activated, and in cell culture, active STAT3 is either required for transformation, enhances transformation, or blocks apoptosis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Human
  • Detection Range:
    0.156 - 10 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEB743HU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.062 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    STAT3
  • Sample Type:
    Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Signal Transducer And Activator Of Transcription 3 (STAT3) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications