Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human SPD ELISA Kit

Human SPD ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEB039HU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEB039HU
Human SPD ELISA Kit
Catalog # MSPP-SEB039HU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human SPD ELISA Kit
Human SPD ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$498.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Human SPD (Surfactant Associated Protein D). The assay range is from 6.25 to 400 ng/ml (Sandwich kit) with a sensitivity of 2.51 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Surfactant protein D (SP-D) belongs to the collectin family.These proteins are oligomeric proteins composed of carbohydrate-recognition domains(CRD)attached to collagenous regions.Collectins are structurally similar to the ficolins although they make use of different CRD structures:C-type lectin domain for the collectins.The lung is the major site of synthesis of SP-D, where the molecules are produced and secreted onto the epithelial surface by alveolar type Ⅱcells and unciliated bronchial epithelial cells. SP-D is also found in different epithelial cells of the gastrointestinal tract and in epithelial cells of various exocrine glands. SP-D is an important factor in the pulmonary anti-microbial defense. The anti-microbial defense mechanisms of SP-D are direct opsonization, neutralization and agglutination.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Human
  • Detection Range:
    6.25 - 400 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEB039HU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    2.51 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    SPD
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Surfactant Associated Protein D (SPD) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.