Order Entry
Puerto Rico
ContactUsLinkComponent
Human SIRT6 ELISA Kit
Human SIRT6 ELISA Kit
Catalog # MSPP-SEE916HU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human SIRT6 ELISA Kit
Catalog # MSPP-SEE916HU
Supplier:  Cloud-Clone
Supplier Number:  SEE916HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    SIRT6
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Sirtuin 6 (SIRT6) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.116 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEE916HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human SIRT6 (Sirtuin 6). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.116 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

SIRT6 encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family.Sirtuins can be divided into 4 main classes in eukaryotes based on amino acid sequence motifs, with yeast Sir2 belonging to class Ia. The deduced 355-amino acid SIRT6 protein is a class IV sirtuin, a class not present in prokaryotes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.