- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:SDC1
- Size:1 kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Syndecan 1 (SDC1) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:1.56 - 100 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.66 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEB966HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human SDC1 (Syndecan 1). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.66 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Syndecan 1 is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-1 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-1 expression has been detected in several different tumor types. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same protein.