Order Entry
Puerto Rico
ContactUsLinkComponent
Human SCF ELISA Kit
Human SCF ELISA Kit
Catalog # MSPP-SEA120HU
CAS Number:  
undefined
Human SCF ELISA Kit
Catalog # MSPP-SEA120HU
Supplier Number:  SEA120HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    SCF
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Stem Cell Factor (SCF) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA120HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human SCF (Stem Cell Factor). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Stem cell factor (SCF;MGF) is a cytokine which binds CD117 (c-Kit). SCF is also known as 'steel factor' or 'c-kit ligand'. SCF exists in two forms, cell surface bound SCF and soluble (or free) SCF. Soluble SCF is produced by the cleavage of surface bound SCF by metalloproteases.SCF is a growth factor important for the survival, proliferation, and differentiation of hematopoietic stem cells and other hematopoietic progenitor cells. One of its roles is to change the BFU-E(burst-forming unit-erythroid) cells, which are the earliest erythrocyte precursors in the erythrocytic series, into the CFU-E (colony-forming unit-erythroid).  SCF, along with bFGF (basic fibroblast growth factor) and LIF (leukemia inhibitory factor), prevents spontaneous differentiation of primitive embryonic stem cells in cell culture.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.